Product Details

SNP ID
rs111934367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69399450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGGCCGGAATCATTCCCTTGGC[C/T]GTTAATTTCTCAGAGGTAAGAAATT
Phenotype
MIM: 601191
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERH PubMed Links

Gene Details

Gene
ERH
Gene Name
enhancer of rudimentary homolog (Drosophila)
There are no transcripts associated with this gene.

Gene
SLC39A9
Gene Name
solute carrier family 39 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252148.1 783 Silent Mutation GCC,GCT A27A NP_001239077.1
NM_001252150.1 783 Silent Mutation GCC,GCT A27A NP_001239079.1
NM_001252151.1 783 Intron NP_001239080.1
NM_001252152.1 783 Intron NP_001239081.1
NM_018375.4 783 Silent Mutation GCC,GCT A27A NP_060845.2
XM_005267834.1 783 Silent Mutation GCC,GCT A27A XP_005267891.1
XM_017021426.1 783 UTR 5 XP_016876915.1

View Full Product Details