Product Details

SNP ID
rs116577677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:62376148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTTTAGTAGTGTGTTTTTCAGC[C/T]GACTTGTAAACTACCTATTTCCCCC
Phenotype
MIM: 607818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC283045 PubMed Links

Gene Details

Gene
LOC283045
Gene Name
uncharacterized LOC283045
There are no transcripts associated with this gene.

Gene
ZNF365
Gene Name
zinc finger protein 365
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014951.2 Intron NP_055766.2
NM_199450.2 Intron NP_955522.1
NM_199451.2 Intron NP_955523.1
NM_199452.3 Intron NP_955524.3
XM_017015937.1 Intron XP_016871426.1
XM_017015938.1 Intron XP_016871427.1
XM_017015939.1 Intron XP_016871428.1
XM_017015940.1 Intron XP_016871429.1
XM_017015941.1 Intron XP_016871430.1

View Full Product Details