Product Details
- SNP ID
-
rs148906362
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:97001275 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGGCACTCCACCCATGACAGGGGG[C/T]GCGTGGTCTGGCAGATGGCATAGCC
- Phenotype
-
MIM: 603742
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLIT1
PubMed Links
Gene Details
- Gene
- SLIT1
- Gene Name
- slit guidance ligand 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003061.2 |
4694 |
Missense Mutation |
CAC,CGC |
H1481R |
NP_003052.2 |
View Full Product Details