Product Details

SNP ID
rs139918773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59831741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTCTTGTGATTAACATTTTCCG[C/T]GATATTGTTGATAGAAGAGACGACA
Phenotype
MIM: 609342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIF PubMed Links

Gene Details

Gene
GIF
Gene Name
gastric intrinsic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005142.2 1163 Missense Mutation ACG,GCG T377A NP_005133.2
XM_011544939.2 1163 Missense Mutation ACG,GCG T363A XP_011543241.1

View Full Product Details