Product Details

SNP ID
rs140833755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:104947164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTTGTGTGATGAAGATAGAGCC[C/T]ATTGTGCTGTCTCTCCAGGACACGT
Phenotype
MIM: 602664
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP4 PubMed Links

Gene Details

Gene
CASP4
Gene Name
caspase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001225.3 1027 Missense Mutation ATA,ATG I318M NP_001216.1
NM_033306.2 1027 Missense Mutation ATA,ATG I262M NP_150649.1
XM_011543019.1 1027 Missense Mutation ATA,ATG I227M XP_011541321.1
XM_017018397.1 1027 Missense Mutation ATA,ATG I227M XP_016873886.1

View Full Product Details