Product Details

SNP ID
rs145558620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55651012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTCTTTCTTCTACATAATCATT[C/T]TTCTGGGAAATCTCCTCATCATGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4S2 PubMed Links

Gene Details

Gene
OR4S2
Gene Name
olfactory receptor family 4 subfamily S member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004059.2 109 Missense Mutation CTT,TTT L37F NP_001004059.2

View Full Product Details