Product Details

SNP ID
rs149645212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65917747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACGTGGGCTCACCTTGGCCAC[C/G]TGAAGCTGGCCTTCAACCACGGCCC
Phenotype
MIM: 602289
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C11orf68 PubMed Links

Gene Details

Gene
C11orf68
Gene Name
chromosome 11 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135635.1 620 Silent Mutation CAC,CAG H198Q NP_001129107.1
NM_031450.3 620 Silent Mutation CAC,CAG H197Q NP_113638.2
Gene
DRAP1
Gene Name
DR1 associated protein 1
There are no transcripts associated with this gene.

View Full Product Details