Product Details

SNP ID
rs150128867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55650904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCTAAGAACTTGACCCATTCC[A/G]TGGAAAAAATAAACAACGTAACTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4S2 PubMed Links

Gene Details

Gene
OR4S2
Gene Name
olfactory receptor family 4 subfamily S member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004059.2 1 Missense Mutation ATG,GTG M1V NP_001004059.2

View Full Product Details