Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001206999.1 | 6035 | Silent Mutation | NP_001193928.1 | ||
NM_007174.2 | 6035 | Silent Mutation | NP_009105.1 | ||
XM_006719206.2 | 6035 | Silent Mutation | XP_006719269.1 | ||
XM_011537783.1 | 6035 | Silent Mutation | XP_011536085.1 | ||
XM_011537784.1 | 6035 | Silent Mutation | XP_011536086.1 | ||
XM_011537785.1 | 6035 | Silent Mutation | XP_011536087.1 | ||
XM_011537787.1 | 6035 | Silent Mutation | XP_011536089.1 | ||
XM_011537788.1 | 6035 | Silent Mutation | XP_011536090.1 | ||
XM_011537789.1 | 6035 | Silent Mutation | XP_011536091.1 | ||
XM_011537790.2 | 6035 | Silent Mutation | XP_011536092.1 | ||
XM_011537791.2 | 6035 | Silent Mutation | XP_011536093.1 | ||
XM_011537792.2 | 6035 | Silent Mutation | XP_011536094.1 | ||
XM_017018735.1 | 6035 | Silent Mutation | XP_016874224.1 | ||
XM_017018736.1 | 6035 | Silent Mutation | XP_016874225.1 | ||
XM_017018737.1 | 6035 | Silent Mutation | XP_016874226.1 | ||
XM_017018738.1 | 6035 | Silent Mutation | XP_016874227.1 | ||
XM_017018739.1 | 6035 | Silent Mutation | XP_016874228.1 |