Product Details
- SNP ID
-
rs144102132
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:119690414 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCTGGAGGCCACGCGCTTGGTGA[C/T]GTGCTCGTTGTACGTGGGTGGGCCT
- Phenotype
-
MIM: 605629
MIM: 602740
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CIT
PubMed Links
Gene Details
- Gene
- CIT
- Gene Name
- citron rho-interacting serine/threonine kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001206999.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1975V |
NP_001193928.1 |
NM_007174.2 |
6003 |
Missense Mutation |
ATC,GTC |
I1933V |
NP_009105.1 |
XM_006719206.2 |
6003 |
Missense Mutation |
ATC,GTC |
I1960V |
XP_006719269.1 |
XM_011537783.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1990V |
XP_011536085.1 |
XM_011537784.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1989V |
XP_011536086.1 |
XM_011537785.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1975V |
XP_011536087.1 |
XM_011537787.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1918V |
XP_011536089.1 |
XM_011537788.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1917V |
XP_011536090.1 |
XM_011537789.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1544V |
XP_011536091.1 |
XM_011537790.2 |
6003 |
Missense Mutation |
ATC,GTC |
I1544V |
XP_011536092.1 |
XM_011537791.2 |
6003 |
Missense Mutation |
ATC,GTC |
I1524V |
XP_011536093.1 |
XM_011537792.2 |
6003 |
Missense Mutation |
ATC,GTC |
I1451V |
XP_011536094.1 |
XM_017018735.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1974V |
XP_016874224.1 |
XM_017018736.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1974V |
XP_016874225.1 |
XM_017018737.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1959V |
XP_016874226.1 |
XM_017018738.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1482V |
XP_016874227.1 |
XM_017018739.1 |
6003 |
Missense Mutation |
ATC,GTC |
I1451V |
XP_016874228.1 |
- Gene
- PRKAB1
- Gene Name
- protein kinase AMP-activated non-catalytic subunit beta 1
There are no transcripts associated with this gene.
View Full Product Details