Product Details

SNP ID
rs148105120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7062569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTAACTATCCTCAGGCATATCCC[A/G]GTGAGGTAGAGAAATCTTGGGACAT
Phenotype
MIM: 120580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1S PubMed Links

Gene Details

Gene
C1S
Gene Name
complement component 1, s subcomponent
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001734.3 463 Missense Mutation AGT,GGT S34G NP_001725.1
NM_201442.2 463 Missense Mutation AGT,GGT S34G NP_958850.1
XM_005253760.1 463 Missense Mutation AGT,GGT S34G XP_005253817.1

View Full Product Details