Product Details

SNP ID
rs150793545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57516866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGCTCGGCGAGTCGCCTCTAC[C/T]TCCCTGGTCAGGCGCTCGATTTCCT
Phenotype
MIM: 126337 MIM: 156560 MIM: 614489
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDIT3 PubMed Links

Gene Details

Gene
DDIT3
Gene Name
DNA damage inducible transcript 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195053.1 790 Silent Mutation GAA,GAG E174E NP_001181982.1
NM_001195054.1 790 Silent Mutation GAA,GAG E174E NP_001181983.1
NM_001195055.1 790 Silent Mutation GAA,GAG E174E NP_001181984.1
NM_001195056.1 790 Silent Mutation GAA,GAG E174E NP_001181985.1
NM_001195057.1 790 Silent Mutation GAA,GAG E151E NP_001181986.1
NM_004083.5 790 Silent Mutation GAA,GAG E151E NP_004074.2
Gene
MARS
Gene Name
methionyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004990.3 790 Intron NP_004981.2
XM_006719398.3 790 Intron XP_006719461.1
Gene
MBD6
Gene Name
methyl-CpG binding domain protein 6
There are no transcripts associated with this gene.

Gene
MIR616
Gene Name
microRNA 616
There are no transcripts associated with this gene.

Gene
MIR6758
Gene Name
microRNA 6758
There are no transcripts associated with this gene.

View Full Product Details