Product Details

SNP ID
rs139794108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20243367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATCACTTCCTGTGTGACATGGA[C/T]CCATTGATGGCTCTATCCTGTGCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR11H4 PubMed Links

Gene Details

Gene
OR11H4
Gene Name
olfactory receptor family 11 subfamily H member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004479.1 576 Silent Mutation GAC,GAT D192D NP_001004479.1

View Full Product Details