Product Details

SNP ID
rs141050108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74716057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAACTTTTCCATAAAAGCCAAA[C/G]TGGACTTAGTGCAGTCAATGATGGA
Phenotype
MIM: 615380
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AREL1 PubMed Links

Gene Details

Gene
AREL1
Gene Name
apoptosis resistant E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.

Gene
FCF1
Gene Name
FCF1 rRNA-processing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318508.1 433 Intron NP_001305437.1
NM_015962.4 433 Intron NP_057046.1
XM_005267730.3 433 UTR 5 XP_005267787.1
XM_011536815.2 433 Intron XP_011535117.1
XM_011536816.2 433 Intron XP_011535118.1

View Full Product Details