Product Details

SNP ID
rs142720326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20242846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAACAGGTCAGCAACACACATC[A/G]TGACAGAGTTTATTCTCCTGGGATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR11H4 PubMed Links

Gene Details

Gene
OR11H4
Gene Name
olfactory receptor family 11 subfamily H member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004479.1 55 Missense Mutation ATG,GTG M19V NP_001004479.1

View Full Product Details