Product Details

SNP ID
rs145941889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20243326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTACATCTCCCAACTCCCCTTC[C/T]GTGGTCCTAATATCATTGATCACTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR11H4 PubMed Links

Gene Details

Gene
OR11H4
Gene Name
olfactory receptor family 11 subfamily H member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004479.1 535 Missense Mutation CGT,TGT R179C NP_001004479.1

View Full Product Details