Product Details
- SNP ID
-
rs146737847
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:60509783 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCCGCGCACCATTTGGGACGGC[A/G]AACAGAAGACACACTGCTTCAAGGA
- Phenotype
-
MIM: 606326
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SIX6
PubMed Links
Gene Details
- Gene
- SIX6
- Gene Name
- SIX homeobox 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007374.2 |
564 |
Missense Mutation |
AAA,GAA |
K129E |
NP_031400.2 |
View Full Product Details