Product Details

SNP ID
rs149623616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75112610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCTTTGCTCCTACCTGCCCAG[C/T]TGTTCTGCAAAAGCCTTGACCCTGC
Phenotype
MIM: 607490 MIM: 612060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDHD PubMed Links

Gene Details

Gene
LDHD
Gene Name
lactate dehydrogenase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153486.3 1333 Silent Mutation CAA,CAG Q450Q NP_705690.2
NM_194436.2 1333 Silent Mutation CAA,CAG Q427Q NP_919417.1
Gene
ZNRF1
Gene Name
zinc and ring finger 1, E3 ubiquitin protein ligase
There are no transcripts associated with this gene.

View Full Product Details