Product Details

SNP ID
rs138610354
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75517568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCACTTCCAGGGCTCCATCCA[C/T]CTCTTCCACCAAGACCTGCCACTGT
Phenotype
MIM: 612185 MIM: 608755
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CASKIN2 PubMed Links
Additional Information
For this assay, SNP(s) [rs56895494] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CASKIN2
Gene Name
CASK interacting protein 2
There are no transcripts associated with this gene.

Gene
LLGL2
Gene Name
LLGL2, scribble cell polarity complex component
There are no transcripts associated with this gene.

Gene
TSEN54
Gene Name
tRNA splicing endonuclease subunit 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207346.2 413 Silent Mutation CAC,CAT H127H NP_997229.2
XM_005257229.3 413 Silent Mutation CAC,CAT H127H XP_005257286.1

View Full Product Details