Product Details

SNP ID
rs148370743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35477632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGCAAAAGACAGAATCGCCAC[C/T]TCATAGAAGATAAAATATCTTCTAT
Phenotype
MIM: 614956
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLFN12L PubMed Links

Gene Details

Gene
SLFN12L
Gene Name
schlafen family member 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195790.1 Intron NP_001182719.1
XM_017024005.1 Intron XP_016879494.1
XM_017024006.1 Intron XP_016879495.1
XM_017024007.1 Intron XP_016879496.1
XM_017024008.1 Intron XP_016879497.1
XM_017024009.1 Intron XP_016879498.1

View Full Product Details