Product Details

SNP ID
rs148859527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76471870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGAAGGCCAGCAGCAGGCCACT[A/G]AGGAAGCCGAAGATGTGGGCGATGT
Phenotype
MIM: 600950 MIM: 614404
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AANAT PubMed Links

Gene Details

Gene
AANAT
Gene Name
aralkylamine N-acetyltransferase
There are no transcripts associated with this gene.

Gene
RHBDF2
Gene Name
rhomboid 5 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005498.3 2989 Silent Mutation CTC,CTT L749L NP_001005498.2
NM_024599.5 2989 Silent Mutation CTC,CTT L778L NP_078875.4
XM_005257669.3 2989 Silent Mutation CTC,CTT L778L XP_005257726.1
XM_005257670.1 2989 Silent Mutation CTC,CTT L749L XP_005257727.1
XM_011525249.2 2989 Silent Mutation CTC,CTT L749L XP_011523551.1
XM_011525250.2 2989 Silent Mutation CTC,CTT L749L XP_011523552.1
XM_011525251.2 2989 Silent Mutation CTC,CTT L749L XP_011523553.1
XM_017025079.1 2989 Silent Mutation CTC,CTT L749L XP_016880568.1

View Full Product Details