Product Details
- SNP ID
-
rs149014968
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40999835 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCCAGTGGGGACACTGCAGCTGC[A/G]AGAGGCACAGCAATCCATGGCAATG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRTAP3-1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs548322379] are located under a probe and SNP(s) [rs9890438] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRTAP3-1
- Gene Name
- keratin associated protein 3-1
There are no transcripts associated with this gene.
- Gene
- KRTAP3-2
- Gene Name
- keratin associated protein 3-2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031959.2 |
52 |
Missense Mutation |
CGC,TGC |
R7C |
NP_114165.1 |
- Gene
- KRTAP3-3
- Gene Name
- keratin associated protein 3-3
There are no transcripts associated with this gene.
View Full Product Details