Product Details
- SNP ID
-
rs150254912
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41347133 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCTCATACTGACTCCTGGTCTC[A/G]TTCAGGACCTGGTTCAGGTCCACAG
- Phenotype
-
MIM: 602761
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT33A
PubMed Links
Gene Details
- Gene
- KRT33A
- Gene Name
- keratin 33A
View Full Product Details