Product Details

SNP ID
rs139869066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18919802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCGTGGAACAATGTCGGCAGCTG[A/G]GTTTGAAGCAGCCCACGCCCGTGCA
Phenotype
MIM: 606942 MIM: 604800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPE PubMed Links

Gene Details

Gene
COPE
Gene Name
coatomer protein complex subunit epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007263.3 128 Intron NP_009194.2
NM_199442.1 128 Intron NP_955474.1
NM_199444.1 128 Intron NP_955476.1
XM_011527655.1 128 Intron XP_011525957.1
XM_011527656.1 128 Intron XP_011525958.1
Gene
DDX49
Gene Name
DEAD-box helicase 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019070.4 128 Missense Mutation AGT,GGT S21G NP_061943.2
XM_011528083.1 128 UTR 5 XP_011526385.1
XM_011528084.2 128 UTR 5 XP_011526386.1
Gene
HOMER3
Gene Name
homer scaffolding protein 3
There are no transcripts associated with this gene.

View Full Product Details