Product Details

SNP ID
rs145580205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35332796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGCAGAAAAGGGTGCAATTCC[G/T]GGGAGACAAGAATAAGAACTGCACA
Phenotype
MIM: 107266
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD22 PubMed Links

Gene Details

Gene
CD22
Gene Name
CD22 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185099.1 368 Missense Mutation CGG,CTG R95L NP_001172028.1
NM_001185100.1 368 Missense Mutation CGG,CTG R95L NP_001172029.1
NM_001185101.1 368 Missense Mutation CGG,CTG R95L NP_001172030.1
NM_001278417.1 368 UTR 5 NP_001265346.1
NM_001771.3 368 Missense Mutation CGG,CTG R95L NP_001762.2

View Full Product Details