Product Details

SNP ID
rs148651180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12319648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATACTCATGTGGTTTGTGTCCA[A/G]AATCAACTCTGATGTGGCTATTAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF563 PubMed Links

Gene Details

Gene
ZNF563
Gene Name
zinc finger protein 563
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145276.2 615 Missense Mutation TCT,TTT S126F NP_660319.1
XM_005259750.4 615 Intron XP_005259807.1
XM_005259751.3 615 Intron XP_005259808.1
XM_006722650.3 615 Intron XP_006722713.1
XM_006722651.3 615 Intron XP_006722714.1
XM_011527698.1 615 Intron XP_011526000.1
XM_011527699.2 615 Missense Mutation TCT,TTT S89F XP_011526001.1
XM_011527700.2 615 Intron XP_011526002.1
XM_017026332.1 615 Intron XP_016881821.1

View Full Product Details