Product Details

SNP ID
rs112305589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54847916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCTCCAGGATTCTGATGAACA[A/G]GACCATCAGGAGGTGTCATACGCAT
Phenotype
MIM: 604955 MIM: 604947
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIR2DS4 PubMed Links

Gene Details

Gene
KIR2DS4
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281971.1 955 UTR 3 NP_001268900.1
NM_001281972.1 955 UTR 3 NP_001268901.1
NM_012314.5 955 Intron NP_036446.3
Gene
KIR3DL2
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2
There are no transcripts associated with this gene.

View Full Product Details