Product Details

SNP ID
rs150045277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12319472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTTGCACTACCATGTGTCTTC[A/G]AAGGTTTCTACGAGAACTGAAGGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF563 PubMed Links

Gene Details

Gene
ZNF563
Gene Name
zinc finger protein 563
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145276.2 791 Nonsense Mutation CGA,TGA R185* NP_660319.1
XM_005259750.4 791 Intron XP_005259807.1
XM_005259751.3 791 Intron XP_005259808.1
XM_006722650.3 791 Intron XP_006722713.1
XM_006722651.3 791 Intron XP_006722714.1
XM_011527698.1 791 Intron XP_011526000.1
XM_011527699.2 791 Nonsense Mutation CGA,TGA R148* XP_011526001.1
XM_011527700.2 791 Intron XP_011526002.1
XM_017026332.1 791 Intron XP_016881821.1

View Full Product Details