Product Details

SNP ID
rs143483780
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:206798917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATGTGCACTGAGGGAGCGTTTC[C/T]GCACAGATCTGCGTGTTCCTTACCA
Phenotype
MIM: 605687
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL19 PubMed Links

Gene Details

Gene
IL19
Gene Name
interleukin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013371.3 48 Intron NP_037503.2
NM_153758.2 48 Missense Mutation CCG,CTG P8L NP_715639.1
XM_011509450.2 48 Intron XP_011507752.1

View Full Product Details