Product Details

SNP ID
rs144631287
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:54139598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCAGTCTTAGGGGTCCCGAGAG[C/T]GGGCAAGGGTGTAGCCAATGGAGAA
Phenotype
MIM: 612320
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDCP2 PubMed Links

Gene Details

Gene
CDCP2
Gene Name
CUB domain containing protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201546.3 1356 Silent Mutation CCA,CCG P424P NP_963840.2

View Full Product Details