Product Details

SNP ID
rs148090964
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:54331940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACGTTCCTGGGAACCAGCCCCA[A/G]AGATTCTGAGTTTGCAGTGCTTTCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C2orf73 PubMed Links

Gene Details

Gene
C2orf73
Gene Name
chromosome 2 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100396.1 Intron NP_001093866.1
XM_005264133.1 Intron XP_005264190.1
XM_005264134.3 Intron XP_005264191.1
XM_011532510.2 Intron XP_011530812.1
XM_011532513.2 Intron XP_011530815.1
XM_017003323.1 Intron XP_016858812.1
XM_017003324.1 Intron XP_016858813.1

View Full Product Details