Product Details

SNP ID
rs151021291
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:54334827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTATGTTTTTCAGACAGCATAAA[A/C]TAGAGGATGCTGCTATAACATATGT
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
C2orf73 PubMed Links
Additional Information
For this assay, SNP(s) [rs141374956] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C2orf73
Gene Name
chromosome 2 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100396.1 149 Missense Mutation ATA,CTA I13L NP_001093866.1
XM_005264133.1 149 Intron XP_005264190.1
XM_005264134.3 149 Intron XP_005264191.1
XM_011532510.2 149 Missense Mutation ATA,CTA I13L XP_011530812.1
XM_011532513.2 149 Intron XP_011530815.1
XM_017003323.1 149 Missense Mutation ATA,CTA I13L XP_016858812.1
XM_017003324.1 149 Missense Mutation ATA,CTA I13L XP_016858813.1

View Full Product Details