Product Details

SNP ID
rs138910107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:127692221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCAGGGTGGGGACGCAGTTCCTG[C/T]CGTGGCTGTCCTTTGAGAGACCCAC
Phenotype
MIM: 609699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGLL PubMed Links

Gene Details

Gene
MGLL
Gene Name
monoglyceride lipase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003794.2 1206 Missense Mutation ACA,GCA T297A NP_001003794.1
NM_001256585.1 1206 Missense Mutation ACA,GCA T277A NP_001243514.1
NM_007283.6 1206 Missense Mutation ACA,GCA T307A NP_009214.1
XM_011512377.1 1206 Missense Mutation ACA,GCA T333A XP_011510679.1
XM_011512378.1 1206 Missense Mutation ACA,GCA T323A XP_011510680.1
XM_011512379.2 1206 Missense Mutation ACA,GCA T297A XP_011510681.1
XM_011512382.1 1206 Missense Mutation ACA,GCA T260A XP_011510684.1
XM_011512383.2 1206 Missense Mutation ACA,GCA T241A XP_011510685.1
XM_017005662.1 1206 Intron XP_016861151.1
XM_017005663.1 1206 Intron XP_016861152.1
XM_017005664.1 1206 Missense Mutation ACA,GCA T271A XP_016861153.1
XM_017005665.1 1206 Missense Mutation ACA,GCA T271A XP_016861154.1
XM_017005666.1 1206 Missense Mutation ACA,GCA T267A XP_016861155.1

View Full Product Details