Product Details

SNP ID
rs144923418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49280775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCCCCATGGCTCCATAATGATT[A/G]GACACGGCAATGAGGTCGTACACAT
Phenotype
MIM: 603486
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf62 PubMed Links

Gene Details

Gene
C3orf62
Gene Name
chromosome 3 open reading frame 62
There are no transcripts associated with this gene.

Gene
MIR4271
Gene Name
microRNA 4271
There are no transcripts associated with this gene.

Gene
USP4
Gene Name
ubiquitin specific peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251877.1 2551 Intron NP_001238806.1
NM_003363.3 2551 Silent Mutation TCC,TCT S871S NP_003354.2
NM_199443.2 2551 Silent Mutation TCC,TCT S824S NP_955475.1

View Full Product Details