Product Details
- SNP ID
-
rs148371622
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139019931 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCGGGAGCGGCGAGCGCGCGTGG[A/G]GACCTGGACTCCCCACGCACGGAGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRR23B
PubMed Links
Gene Details
- Gene
- PRR23B
- Gene Name
- proline rich 23B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001013650.2 |
996 |
Missense Mutation |
CCC,CTC |
P244L |
NP_001013672.1 |
View Full Product Details