Product Details

SNP ID
rs140656046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:22388009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTGTAGGCTAAATAAGCTCTT[C/T]GGCTCCTCGAGTGTCCTTCGTTATT
Phenotype
MIM: 612303
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRA3 PubMed Links

Gene Details

Gene
ADGRA3
Gene Name
adhesion G protein-coupled receptor A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145290.3 3932 Missense Mutation CAA,CGA Q1221R NP_660333.2
XM_005248137.4 3932 Intron XP_005248194.1
XM_011513811.2 3932 Missense Mutation CAA,CGA Q995R XP_011512113.1
XM_017007834.1 3932 Missense Mutation CAA,CGA Q1024R XP_016863323.1

View Full Product Details