Product Details

SNP ID
rs141591864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185407971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTTTTGACGTTATACCGATCAA[C/T]TGGTTTAGTACCAGCTGCACCTCAA
Phenotype
MIM: 611482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD37 PubMed Links

Gene Details

Gene
ANKRD37
Gene Name
ankyrin repeat domain 37
There are no transcripts associated with this gene.

Gene
C4orf47
Gene Name
chromosome 4 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114357.1 1203 Intron NP_001107829.1
XM_005263019.3 1203 Intron XP_005263076.1
XM_011531989.2 1203 Intron XP_011530291.1
XM_017008236.1 1203 Intron XP_016863725.1
XM_017008237.1 1203 Intron XP_016863726.1
XM_017008238.1 1203 Intron XP_016863727.1
XM_017008239.1 1203 Intron XP_016863728.1
XM_017008240.1 1203 Intron XP_016863729.1
XM_017008241.1 1203 Intron XP_016863730.1
XM_017008242.1 1203 Intron XP_016863731.1
Gene
UFSP2
Gene Name
UFM1 specific peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018359.3 1203 Silent Mutation CAA,CAG Q362Q NP_060829.2

View Full Product Details