Product Details

SNP ID
rs141674956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115962926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTACGCCTGCCGCCCTGGCTCGT[A/G]CCGCTGCACTACGATCTGGAGCTGT
Phenotype
MIM: 610046
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LVRN PubMed Links

Gene Details

Gene
LVRN
Gene Name
laeverin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173800.4 487 Silent Mutation GTA,GTG V103V NP_776161.3
XM_011543263.2 487 Silent Mutation GTA,GTG V103V XP_011541565.1
XM_011543264.2 487 Silent Mutation GTA,GTG V103V XP_011541566.1
XM_017009215.1 487 Silent Mutation GTA,GTG V103V XP_016864704.1

View Full Product Details