Product Details
- SNP ID
-
rs147116160
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:73123630 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTCCCGGGCGCAACTTCAGCTCC[G/T]TGCAGGGCTGCAGAGAGGTCAGCAG
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105379030
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs637450] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105379030
- Gene Name
- uncharacterized LOC105379030
There are no transcripts associated with this gene.
- Gene
- TMEM171
- Gene Name
- transmembrane protein 171
View Full Product Details