Product Details

SNP ID
rs151305324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73123606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTTGGGGCTGTGATCCTGGCCC[A/G]CTCCCGGGCGCAACTTCAGCTCCGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105379030 PubMed Links

Gene Details

Gene
LOC105379030
Gene Name
uncharacterized LOC105379030
There are no transcripts associated with this gene.

Gene
TMEM171
Gene Name
transmembrane protein 171
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161342.2 437 Missense Mutation CAC,CGC H78R NP_001154814.1
NM_173490.7 437 Missense Mutation CAC,CGC H78R NP_775761.4
XM_011543156.1 437 Missense Mutation CAC,CGC H78R XP_011541458.1

View Full Product Details