Product Details

SNP ID
rs116953262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:120541140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCTGTGCAAGTGAAGTCATAT[A/C]CTTTTTCATTATGGCAGGCAGAAAC
Phenotype
MIM: 600026
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNTB1 PubMed Links

Gene Details

Gene
SNTB1
Gene Name
syntrophin beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021021.3 Intron NP_066301.1
XM_011517239.1 Intron XP_011515541.1

View Full Product Details