Product Details

SNP ID
rs141348898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153456286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTCCGTTCTAAGCGCGTGCAA[C/T]TTGGCAGCACTCTCCTGCAGCTGCC
Phenotype
MIM: 300540 MIM: 300370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAUS7 PubMed Links

Gene Details

Gene
HAUS7
Gene Name
HAUS augmin like complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017518.7 758 Silent Mutation AAA,AAG K238K NP_059988.3
Gene
TREX2
Gene Name
three prime repair exonuclease 2
There are no transcripts associated with this gene.

View Full Product Details