Product Details

SNP ID
rs145111887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100914440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGACACAATGGAGGTAGTCTAC[C/T]ACATTATGGGTGAAGAGTGAGCGCA
Phenotype
MIM: 300684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XKRX PubMed Links

Gene Details

Gene
XKRX
Gene Name
XK related, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_212559.2 1221 Silent Mutation GTA,GTG V416V NP_997724.2
XM_005262130.2 1221 Silent Mutation GTA,GTG V212V XP_005262187.1
XM_011530954.2 1221 Intron XP_011529256.1
XM_011530955.1 1221 Silent Mutation GTA,GTG V300V XP_011529257.1
XM_017029517.1 1221 Silent Mutation GTA,GTG V212V XP_016885006.1

View Full Product Details