Product Details

SNP ID
rs1131872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6611971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCCCTCACCCGCCCCACGCCG[T/C]TCAGCTGGCTCCCCGGCCCGCGCCG
Phenotype
MIM: 603057 MIM: 602366 MIM: 615818 MIM: 600475 MIM: 607998
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
DCHS1 PubMed Links

Gene Details

Gene
DCHS1
Gene Name
dachsous cadherin-related 1
There are no transcripts associated with this gene.

Gene
ILK
Gene Name
integrin linked kinase
There are no transcripts associated with this gene.

Gene
RRP8
Gene Name
ribosomal RNA processing 8, methyltransferase, homolog (yeast)
There are no transcripts associated with this gene.

Gene
TAF10
Gene Name
TATA-box binding protein associated factor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006284.3 274 Silent Mutation GAA,GAG E73E NP_006275.1
Gene
TPP1
Gene Name
tripeptidyl peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000391.3 274 Intron NP_000382.3

View Full Product Details