Product Details

SNP ID
rs181377829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:715354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGCCTCCTGGCCCCGGCTGCCC[A/G]CGCCGGCTACTCCGAGGAGCGCTGC
Phenotype
MIM: 614666 MIM: 609082 MIM: 610998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC78 PubMed Links

Gene Details

Gene
CCDC78
Gene Name
coiled-coil domain containing 78
There are no transcripts associated with this gene.

Gene
FAM173A
Gene Name
family with sequence similarity 173 member A
There are no transcripts associated with this gene.

Gene
FBXL16
Gene Name
F-box and leucine rich repeat protein 16
There are no transcripts associated with this gene.

Gene
METRN
Gene Name
meteorin, glial cell differentiation regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024042.3 182 Missense Mutation CAC,CGC H22R NP_076947.1

View Full Product Details