Product Details

SNP ID
rs181393981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127709546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCGCACGCTCAACCAGCAGGTG[C/G]ATGAGATCACAGACCTCAACGAGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP157 PubMed Links

Gene Details

Gene
CFAP157
Gene Name
cilia and flagella associated protein 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012502.2 329 Missense Mutation CAT,GAT H96D NP_001012520.2
XM_006717064.3 329 Missense Mutation CAT,GAT H191D XP_006717127.2
XM_011518559.2 329 Missense Mutation CAT,GAT H191D XP_011516861.2
XM_017014629.1 329 Missense Mutation CAT,GAT H191D XP_016870118.1
XM_017014630.1 329 UTR 5 XP_016870119.1
Gene
PTRH1
Gene Name
peptidyl-tRNA hydrolase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002913.1 329 Intron NP_001002913.1
XM_006716955.3 329 Intron XP_006717018.1
XM_017014276.1 329 Intron XP_016869765.1
XM_017014277.1 329 Intron XP_016869766.1
XM_017014278.1 329 Intron XP_016869767.1
Gene
TTC16
Gene Name
tetratricopeptide repeat domain 16
There are no transcripts associated with this gene.

View Full Product Details