Product Details

SNP ID
rs181899139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18592150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGATCAGCGAGGTTCTCCAGGA[A/C]CTTAGGTTTGATGCGGAATCTGCCG
Phenotype
MIM: 604237
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C19orf60 PubMed Links

Gene Details

Gene
C19orf60
Gene Name
chromosome 19 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100418.1 614 Missense Mutation GAA,GAC E192D NP_001093888.1
NM_001100419.1 614 Missense Mutation GAA,GAC E170D NP_001093889.1
XM_005259977.1 614 Missense Mutation GAA,GAC E204D XP_005260034.1
Gene
CRLF1
Gene Name
cytokine receptor like factor 1
There are no transcripts associated with this gene.

View Full Product Details