Product Details

SNP ID
rs184963094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:77830760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGCAATATCAACTCTAATCATT[C/T]CAGGTATTGGTAATCACTGTATAAA
Phenotype
MIM: 300715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAGT1 PubMed Links

Gene Details

Gene
MAGT1
Gene Name
magnesium transporter 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032121.5 Intron NP_115497.4

View Full Product Details