Product Details
- SNP ID
-
hCV182082849
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:77857375 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTTACATTGACATCAGTTCTGTC[G/T]GCGATCCACCGGGCAATCTGCTCAG
- Phenotype
-
MIM: 300715
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MAGT1
PubMed Links
Gene Details
- Gene
- MAGT1
- Gene Name
- magnesium transporter 1
There are no transcripts associated with this gene.
View Full Product Details