Product Details
- SNP ID
-
rs191626545
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49323338 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGGAAGCTGGGTAGGCCCTGAGG[A/G]GCCTCGGTAAGGTAAGGCACGGGGG
- Phenotype
-
MIM: 615229
MIM: 603872
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1QL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112986873] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1QL4
- Gene Name
- complement C1q like 4
There are no transcripts associated with this gene.
- Gene
- LOC101927267
- Gene Name
- uncharacterized LOC101927267
There are no transcripts associated with this gene.
- Gene
- TROAP
- Gene Name
- trophinin associated protein
View Full Product Details